Learn More
ABclonal Technology VSNL1/VILIP-1 Rabbit mAb
SDP
Catalog No. 5000007329
Encompass_Preferred
This gene is a member of the visinin recoverin subfamily of neuronal calcium sensor proteins The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase Alternatively spliced transcript variants have been observed but their full-length nature has not been determined
Catalog No. 50-000-07329 Supplier ABclonal Technology Supplier No. A2210020UL
This gene is a member of the visinin recoverin subfamily of neuronal calcium sensor proteins The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase Alternatively spliced transcript variants have been observed but their full-length nature has not been determined