Learn More
Aviva Systems Biology Corp VSTM2A Antibody - middle region (ARP55813_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55813_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VSTM2A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human VSTM2A. Alternate names: MGC33530, VSTM2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61623