Learn More
AdipoGen VSTM5 msFc ms non-lytic
SDP
Catalog No. 502562821
Encompass_Preferred
Recombinant protein. The extracellular domain of mouse VSTM5 aa 28-146 is fused to the N-terminus of the mouse Fc IgG2a non-lytic mutant. Source CHO cells. Lyophilized. Contains PBS. The V-set and transmembrane domain-containing protein VSTM family is composed of 8 proteins VSTM1v1, VSTM1v2, VSTM2a, VSTM2b, VSTM2L, VSTM3, VSTM4 and VSTM 5. Members of the VSTM family are associated with the regulation of functions of immune cells, adipose cells and neuronal cells. The VSTM family is an immunoglobulin Ig superfamily that shares structural similarities with the B7-like transmembrane proteins involved in immune regulation. VSTM1 and VSTM4 play positive and negative roles in macrophage activation, respectively. VSTM3 TIGIT acts as a co-inhibitory receptor that suppresses the cytokine production of T cells and NK cells.
Catalog No. 50-256-2821 Supplier AdipoGen Supplier No. AG40B0239C050
Recombinant protein. The extracellular domain of mouse VSTM5 aa 28-146 is fused to the N-terminus of the mouse Fc IgG2a non-lytic mutant. Source CHO cells. Lyophilized. Contains PBS. The V-set and transmembrane domain-containing protein VSTM family is composed of 8 proteins VSTM1v1, VSTM1v2, VSTM2a, VSTM2b, VSTM2L, VSTM3, VSTM4 and VSTM 5. Members of the VSTM family are associated with the regulation of functions of immune cells, adipose cells and neuronal cells. The VSTM family is an immunoglobulin Ig superfamily that shares structural similarities with the B7-like transmembrane proteins involved in immune regulation. VSTM1 and VSTM4 play positive and negative roles in macrophage activation, respectively. VSTM3 TIGIT acts as a co-inhibitory receptor that suppresses the cytokine production of T cells and NK cells.