Learn More
Aviva Systems Biology Corp VSX1 Antibody - N-terminal region (ARP39108_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39108_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against VSX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VSX1. Alternate names: PPD, KTCN, PPCD, RINX, KTCN1, CAASDSApplicationWB. Species reactivity: Cow, Human
Catalog No. 50-014-53125