Learn More
Aviva Systems Biology Corp VSX1 Antibody - middle region (ARP35882_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35882_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VSX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human VSX1. Alternate names: KTCN, PPCD, PPD, RINX, KTCN1, CAASDSApplicationWB. Species reactivity: Human
Catalog No. 50-014-55907