Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Steel, 17.40 mm, 1 ea
Catalog No. 5000547870
Encompass
This metric pin gage is a high-precision tool designed for accurate hole diameter measurement and calibration. Manufactured to Class ZZ standards from durable steel, this gage provides reliable performance in industrial inspection and quality control applications.
  • Class ZZ tolerance of 0.005 mm
  • Manufactured from steel with a hardness of 60/65 Rc
  • Surface finish of 10 microinch
  • Roundness within 0.0001 in.
  • Includes a certificate of accuracy
  • NIST traceable
  • Individual gage member
Catalog No. 50-005-47870 Supplier MSC Supplier No. 68108455
This metric pin gage is a high-precision tool designed for accurate hole diameter measurement and calibration. Manufactured to Class ZZ standards from durable steel, this gage provides reliable performance in industrial inspection and quality control applications.
  • Class ZZ tolerance of 0.005 mm
  • Manufactured from steel with a hardness of 60/65 Rc
  • Surface finish of 10 microinch
  • Roundness within 0.0001 in.
  • Includes a certificate of accuracy
  • NIST traceable
  • Individual gage member