Learn More
MSC Vermont Gage Pin Gage, 13.59 mm, Class ZZ, Minus, 1 ea
Catalog No. 5000511833
Encompass
This metric pin gage is designed for precise measurement of hole sizes. It is manufactured from durable tool steel with a high-quality finish for reliable performance in various inspection and quality control applications.
  • Class ZZ accuracy
  • Minus tolerance of +0.000/-0.005
  • Constructed from tool steel with a 10 microfinish or better
  • Includes certificate of accuracy with NIST traceability
  • Overall length of 2 in.
Catalog No. 50-005-11833 Supplier MSC Supplier No. 40441040
This metric pin gage is designed for precise measurement of hole sizes. It is manufactured from durable tool steel with a high-quality finish for reliable performance in various inspection and quality control applications.
  • Class ZZ accuracy
  • Minus tolerance of +0.000/-0.005
  • Constructed from tool steel with a 10 microfinish or better
  • Includes certificate of accuracy with NIST traceability
  • Overall length of 2 in.