Learn More
MSC Vermont Gage Class ZZ Minus Plug Gage, 7.61 mm, 2 in. OAL, 1 ea.
Catalog No. 5000511600
Encompass
This Class ZZ minus plug gage is designed for precise metrology applications, including NoGo inspection and the calibration of micrometers, verniers, and other measurement devices. Manufactured in the USA, this tool is constructed from high-quality steel and features a 10 microfinish surface for enhanced durability and reliable performance.
  • Diameter of 7.61 mm
  • Overall length of 2 in.
  • Tolerance of 0.00508 mm
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-11600 Supplier MSC Supplier No. 40435067
This Class ZZ minus plug gage is designed for precise metrology applications, including NoGo inspection and the calibration of micrometers, verniers, and other measurement devices. Manufactured in the USA, this tool is constructed from high-quality steel and features a 10 microfinish surface for enhanced durability and reliable performance.
  • Diameter of 7.61 mm
  • Overall length of 2 in.
  • Tolerance of 0.00508 mm
  • Includes a certificate of accuracy with NIST traceability