Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 15.92 mm, 1 ea
Catalog No. 5000511290
Encompass
This Class ZZ Plus pin gage is designed for precision inspection tasks, including verifying hole sizes, setting micrometers, and calibrating verniers and other measuring devices. Constructed from durable tool steel with a hardness of 60/65 Rc, this gage features a 15.92 mm diameter and an overall length of 2.0000 in.
  • Tolerance of 0.005 mm (0.0002 in.)
  • Go (Plus) configuration
  • Steel finish with 10 microinch or better surface quality
  • Roundness within 0.0001 in.
  • Complies with ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-11290 Supplier MSC Supplier No. 68106970
This Class ZZ Plus pin gage is designed for precision inspection tasks, including verifying hole sizes, setting micrometers, and calibrating verniers and other measuring devices. Constructed from durable tool steel with a hardness of 60/65 Rc, this gage features a 15.92 mm diameter and an overall length of 2.0000 in.
  • Tolerance of 0.005 mm (0.0002 in.)
  • Go (Plus) configuration
  • Steel finish with 10 microinch or better surface quality
  • Roundness within 0.0001 in.
  • Complies with ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability