Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, Tool Steel, 18.15 mm Diameter, 2 in. Length, 1 ea.
Catalog No. 5000511291
Encompass
This Class ZZ Plus plug gage is designed for precision inspection tasks and measuring applications. Constructed from durable tool steel, it provides reliable performance for go inspection, setting micrometers, or calibrating verniers and other measuring devices. This gage adheres to ASME B89.1 standards and ensures high measurement accuracy in demanding industrial environments.
  • Manufactured from high-quality tool steel
  • Features a 10 microinch finish for smooth operation
  • Includes a certificate of accuracy with NIST traceability
  • Measures 18.15 mm in diameter with a 2.0000 in. overall length
  • Tolerance of 0.005 mm
  • Marked with size and direction of tolerance for easy identification
Catalog No. 50-005-11291 Supplier MSC Supplier No. 68109206
This Class ZZ Plus plug gage is designed for precision inspection tasks and measuring applications. Constructed from durable tool steel, it provides reliable performance for go inspection, setting micrometers, or calibrating verniers and other measuring devices. This gage adheres to ASME B89.1 standards and ensures high measurement accuracy in demanding industrial environments.
  • Manufactured from high-quality tool steel
  • Features a 10 microinch finish for smooth operation
  • Includes a certificate of accuracy with NIST traceability
  • Measures 18.15 mm in diameter with a 2.0000 in. overall length
  • Tolerance of 0.005 mm
  • Marked with size and direction of tolerance for easy identification