Learn More
MSC Vermont Gage Pin Gage, 7.74 mm, Tool Steel, 1 ea.
Catalog No. 5000511141
Encompass
This Class ZZ pin gage is designed for precise measurement of hole sizes and features a diameter of 7.74 mm with a 2.0000 in. overall length. Manufactured from durable 52100 tool steel, this gage is heat-treated to 60 to 65 Rc for long-lasting wear resistance in industrial applications.
  • Manufactured from 52100 tool steel
  • Heat-treated to 60 to 65 Rc hardness
  • Finish of 10 microinch or better
  • Roundness within 0.0001 in. (0.0025 mm)
  • Tolerance of 0.0002 in. (0.005 mm)
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-11141 Supplier MSC Supplier No. 68098797
This Class ZZ pin gage is designed for precise measurement of hole sizes and features a diameter of 7.74 mm with a 2.0000 in. overall length. Manufactured from durable 52100 tool steel, this gage is heat-treated to 60 to 65 Rc for long-lasting wear resistance in industrial applications.
  • Manufactured from 52100 tool steel
  • Heat-treated to 60 to 65 Rc hardness
  • Finish of 10 microinch or better
  • Roundness within 0.0001 in. (0.0025 mm)
  • Tolerance of 0.0002 in. (0.005 mm)
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards