Learn More
MSC Vermont Gage Plug Gage, Tool Steel, 5.62 mm, 2 in. Length, 1 ea.
Catalog No. 5000510654
Encompass
This Class ZZ plus plug gage is designed for high-precision hole size inspection and for setting measuring devices such as micrometers and verniers. Constructed from 52100 tool steel and heat-treated to 60–62 RC, this gage ensures durability and wear resistance in demanding measurement applications. It is manufactured to ASME B89.1 standards and includes a certificate of accuracy with NIST traceability to ensure consistent and reliable performance.
  • Manufactured from 52100 tool steel with a 10 micro-finish or better
  • Heat-treated to 60–62 RC for enhanced durability
  • Meets ASME B89.1 standards for quality assurance
  • Includes a certificate of accuracy with NIST traceability
  • Roundness within 0.0001 in.
  • Overall length of 2.0000 in.
Catalog No. 50-005-10654 Supplier MSC Supplier No. 68096676
This Class ZZ plus plug gage is designed for high-precision hole size inspection and for setting measuring devices such as micrometers and verniers. Constructed from 52100 tool steel and heat-treated to 60–62 RC, this gage ensures durability and wear resistance in demanding measurement applications. It is manufactured to ASME B89.1 standards and includes a certificate of accuracy with NIST traceability to ensure consistent and reliable performance.
  • Manufactured from 52100 tool steel with a 10 micro-finish or better
  • Heat-treated to 60–62 RC for enhanced durability
  • Meets ASME B89.1 standards for quality assurance
  • Includes a certificate of accuracy with NIST traceability
  • Roundness within 0.0001 in.
  • Overall length of 2.0000 in.