Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Tool Steel, 17.77 mm, 1 ea.
Catalog No. 5000510391
Encompass
This Class ZZ pin gage is designed for precision Go inspection, checking micrometers, verniers, and other measurement devices. Constructed from durable tool steel with a hardness of 60/65 Rc, this gage features a 17.77 mm diameter and is built to comply with ASME B89.1 standards.
  • Reversible style for extended service life
  • Surface finish of 10 microinch or better
  • Plus tolerance direction for Go applications
  • Includes a certificate of accuracy with NIST traceability
  • Marked with size and direction of tolerance
  • Roundness within 0.0001 in. (0.0025 mm)
  • Tolerance of 0.00508 mm
Catalog No. 50-005-10391 Supplier MSC Supplier No. 68108828
This Class ZZ pin gage is designed for precision Go inspection, checking micrometers, verniers, and other measurement devices. Constructed from durable tool steel with a hardness of 60/65 Rc, this gage features a 17.77 mm diameter and is built to comply with ASME B89.1 standards.
  • Reversible style for extended service life
  • Surface finish of 10 microinch or better
  • Plus tolerance direction for Go applications
  • Includes a certificate of accuracy with NIST traceability
  • Marked with size and direction of tolerance
  • Roundness within 0.0001 in. (0.0025 mm)
  • Tolerance of 0.00508 mm