Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 8.41 mm, 2 in., 1 ea.
Catalog No. 5000510352
Encompass
This Class ZZ plus plug gage is a precision tool designed for inspecting hole sizes, setting micrometers, and calibrating verniers or other measuring devices. Constructed from high-quality tool steel, the gage offers durability and high precision for critical measurement applications.
  • Features a nominal diameter of 8.41 mm and an overall length of 2.0000 in.
  • Manufactured from tool steel with a hardness of 60 to 65 HRC.
  • Includes a certificate of accuracy with NIST traceability to ensure reliable performance.
  • Built to ASME B89.1 standards for quality assurance.
  • Designed with a 10 microfinish or better for smooth surface quality.
  • Reversible style allows for extended product life.
Catalog No. 50-005-10352 Supplier MSC Supplier No. 68099464
This Class ZZ plus plug gage is a precision tool designed for inspecting hole sizes, setting micrometers, and calibrating verniers or other measuring devices. Constructed from high-quality tool steel, the gage offers durability and high precision for critical measurement applications.
  • Features a nominal diameter of 8.41 mm and an overall length of 2.0000 in.
  • Manufactured from tool steel with a hardness of 60 to 65 HRC.
  • Includes a certificate of accuracy with NIST traceability to ensure reliable performance.
  • Built to ASME B89.1 standards for quality assurance.
  • Designed with a 10 microfinish or better for smooth surface quality.
  • Reversible style allows for extended product life.