Learn More
MSC Vermont Gage Pin Gage, Steel, 18.99 mm, Class ZZ Minus, 2 in. OAL, 1 ea.
Catalog No. 5000510200
Encompass
This Class ZZ minus plug and pin gage is a precision measurement tool designed for checking hole sizes, setting micrometers, and performing NoGo inspections on verniers and other measuring instruments. Constructed from durable steel, it features a 18.99 mm diameter and a 2 in. overall length.
  • Manufactured to a tolerance of 0.005 mm
  • Includes a certificate of accuracy with NIST traceability
  • Surface finish of 10 microinch or better
  • Hardness rating of 60 to 65 Rc
  • Round within 0.0025 mm
  • Marked with size and direction of tolerance for identification
Catalog No. 50-005-10200 Supplier MSC Supplier No. 40446445
This Class ZZ minus plug and pin gage is a precision measurement tool designed for checking hole sizes, setting micrometers, and performing NoGo inspections on verniers and other measuring instruments. Constructed from durable steel, it features a 18.99 mm diameter and a 2 in. overall length.
  • Manufactured to a tolerance of 0.005 mm
  • Includes a certificate of accuracy with NIST traceability
  • Surface finish of 10 microinch or better
  • Hardness rating of 60 to 65 Rc
  • Round within 0.0025 mm
  • Marked with size and direction of tolerance for identification