Learn More
MSC Vermont Gage Plug And Pin Gage, Class ZZ, Minus, 1.86 mm, 2 in., 1 ea
Catalog No. 5000516707
Encompass
This Class ZZ minus plug and pin gage is a precision measurement tool designed for checking hole sizes, setting micrometers, and verifying the accuracy of verniers and other measuring devices. It is suitable for NoGo inspections and provides reliable, repeatable measurements in various industrial applications.
  • Manufactured from durable tool steel for extended service life
  • Features a 1.86 mm diameter and 2 in. overall length
  • Includes a certificate of accuracy with NIST traceability
  • Constructed as a reversible style for versatile use
  • Provides a tolerance of 0.00508 mm
  • Surface finish of 10 micro
Catalog No. 50-005-16707 Supplier MSC Supplier No. 40429318
This Class ZZ minus plug and pin gage is a precision measurement tool designed for checking hole sizes, setting micrometers, and verifying the accuracy of verniers and other measuring devices. It is suitable for NoGo inspections and provides reliable, repeatable measurements in various industrial applications.
  • Manufactured from durable tool steel for extended service life
  • Features a 1.86 mm diameter and 2 in. overall length
  • Includes a certificate of accuracy with NIST traceability
  • Constructed as a reversible style for versatile use
  • Provides a tolerance of 0.00508 mm
  • Surface finish of 10 micro