Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Tool Steel, 22.38 mm, 2 in., 1 ea.
Catalog No. 5000516582
Encompass
This Class ZZ pin gage is designed for precision inspection and measurement tasks, including setting micrometers, verniers, and other measuring devices. Engineered with a 22.38 mm diameter, this tool provides reliable NoGo inspection capabilities for checking hole sizes within specified tolerances.
  • Class ZZ tolerance for accurate measurement
  • Constructed from durable tool steel
  • Features a 10 microfinish or better for surface quality
  • Includes a certificate of accuracy with NIST traceability
  • Overall length of 2 in.
  • Reversible style for extended service life
Catalog No. 50-005-16582 Supplier MSC Supplier No. 40449837
This Class ZZ pin gage is designed for precision inspection and measurement tasks, including setting micrometers, verniers, and other measuring devices. Engineered with a 22.38 mm diameter, this tool provides reliable NoGo inspection capabilities for checking hole sizes within specified tolerances.
  • Class ZZ tolerance for accurate measurement
  • Constructed from durable tool steel
  • Features a 10 microfinish or better for surface quality
  • Includes a certificate of accuracy with NIST traceability
  • Overall length of 2 in.
  • Reversible style for extended service life