Learn More
MSC Vermont Gage Pin Gage, 7.47 mm Diameter, Class ZZ Minus, Steel, 2 in. OAL, 1 ea.
Catalog No. 5000516467
Encompass
This Class ZZ minus plug and pin gage is a precision instrument designed for accurate hole diameter inspection and calibration of measuring devices. Constructed from durable steel, it is suited for applications such as setting micrometers and performing no-go inspections. This reversible pin gage offers consistent measurement accuracy and includes a certificate of accuracy with NIST traceability to ensure quality standards.
  • Diameter of 7.47 mm
  • Overall length of 5.08 cm (2 in.)
  • Class ZZ minus tolerance of 0.00508 mm
  • Steel material with a 10 micro finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-16467 Supplier MSC Supplier No. 40434920
This Class ZZ minus plug and pin gage is a precision instrument designed for accurate hole diameter inspection and calibration of measuring devices. Constructed from durable steel, it is suited for applications such as setting micrometers and performing no-go inspections. This reversible pin gage offers consistent measurement accuracy and includes a certificate of accuracy with NIST traceability to ensure quality standards.
  • Diameter of 7.47 mm
  • Overall length of 5.08 cm (2 in.)
  • Class ZZ minus tolerance of 0.00508 mm
  • Steel material with a 10 micro finish
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, verniers, and other measuring devices