Learn More
MSC Vermont Gage Plug Gage, Tool Steel, 3.61 mm Diameter, 2.0000 in. Length, 1 ea
Catalog No. 5000516074
Encompass
This Class ZZ Plus plug gage is designed for precise Go inspection, setting micrometers, and calibrating verniers or other measuring instruments. Constructed from tool steel, it offers reliable performance and durability in industrial inspection applications.
  • Manufactured from durable tool steel
  • Measures 3.61 mm in diameter with a 2.0000 in. overall length
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish
  • Complies with ASME B89.1 standards
  • Maintains a tolerance of 0.00508 mm
Catalog No. 50-005-16074 Supplier MSC Supplier No. 68094663
This Class ZZ Plus plug gage is designed for precise Go inspection, setting micrometers, and calibrating verniers or other measuring instruments. Constructed from tool steel, it offers reliable performance and durability in industrial inspection applications.
  • Manufactured from durable tool steel
  • Measures 3.61 mm in diameter with a 2.0000 in. overall length
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish
  • Complies with ASME B89.1 standards
  • Maintains a tolerance of 0.00508 mm