Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 4.78 mm, 2.0000 in. OAL, 1 ea.
Catalog No. 5000516078
Encompass
This Class ZZ pin gage is a precision-engineered tool designed for high-accuracy inspection tasks. It is ideal for go/no-go inspection, setting micrometers, and calibrating verniers or other measuring instruments. Constructed from durable 52100 tool steel, this gage features a plus tolerance and is manufactured to meet rigorous industry standards.
  • Constructed from 52100 tool steel for durability
  • Heat-treated to a hardness of 60 to 62 RC
  • Finish of 10 microinch or better for smooth surface quality
  • Round within 0.0001 in. (0.0025 mm)
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Marked with size and direction of tolerance
Catalog No. 50-005-16078 Supplier MSC Supplier No. 68095835
This Class ZZ pin gage is a precision-engineered tool designed for high-accuracy inspection tasks. It is ideal for go/no-go inspection, setting micrometers, and calibrating verniers or other measuring instruments. Constructed from durable 52100 tool steel, this gage features a plus tolerance and is manufactured to meet rigorous industry standards.
  • Constructed from 52100 tool steel for durability
  • Heat-treated to a hardness of 60 to 62 RC
  • Finish of 10 microinch or better for smooth surface quality
  • Round within 0.0001 in. (0.0025 mm)
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Marked with size and direction of tolerance