Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 3.14 mm Diameter, 2.0000 in. Length, Tool Steel, 1 ea.
Catalog No. 5000514683
Encompass
This Class ZZ Plus plug gage is designed for precise metrology applications, providing reliable diameter measurement for inspection and calibration tasks. Manufactured from high-quality tool steel, it ensures durability and consistent performance in demanding environments.
  • Diameter of 3.14 mm
  • Overall length of 2.0000 in.
  • Tolerance of 0.00508 mm
  • Finish of 10 micro
  • Manufactured to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-14683 Supplier MSC Supplier No. 68094192
This Class ZZ Plus plug gage is designed for precise metrology applications, providing reliable diameter measurement for inspection and calibration tasks. Manufactured from high-quality tool steel, it ensures durability and consistent performance in demanding environments.
  • Diameter of 3.14 mm
  • Overall length of 2.0000 in.
  • Tolerance of 0.00508 mm
  • Finish of 10 micro
  • Manufactured to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices