Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 5.31 mm, 2.0000 in. OAL, Tool Steel, 1 ea.
Catalog No. 5000514687
Encompass
This Class ZZ Plus plug gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. It features a nominal diameter of 5.31 mm and an overall length of 2.0000 in.
  • Constructed from tool steel for durability
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Finish of 10 micro
  • Tolerance of 0.00508 mm
  • Made in the United States
Catalog No. 50-005-14687 Supplier MSC Supplier No. 68096361
This Class ZZ Plus plug gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. It features a nominal diameter of 5.31 mm and an overall length of 2.0000 in.
  • Constructed from tool steel for durability
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Finish of 10 micro
  • Tolerance of 0.00508 mm
  • Made in the United States