Learn More
MSC Vermont Gage Class ZZ Pin Gage, 13.95 mm, 2 in. Length, 1 ea.
Catalog No. 5000514587
Encompass
This Class ZZ minus plug and pin gage is a precision measurement tool with a diameter of 13.95 mm and an overall length of 2 in. Designed for high-accuracy inspection, it is manufactured from durable tool steel with a hardness of 60/65 Rc to ensure long-term performance in various metrology applications.
  • Hardened tool steel construction for wear resistance
  • Features a 10 microinch or better surface finish
  • Includes a certificate of accuracy with NIST traceability
  • Marked with diameter and direction of tolerance for easy identification
  • Suitable for setting micrometers, verniers, and other measuring devices
Catalog No. 50-005-14587 Supplier MSC Supplier No. 40441404
This Class ZZ minus plug and pin gage is a precision measurement tool with a diameter of 13.95 mm and an overall length of 2 in. Designed for high-accuracy inspection, it is manufactured from durable tool steel with a hardness of 60/65 Rc to ensure long-term performance in various metrology applications.
  • Hardened tool steel construction for wear resistance
  • Features a 10 microinch or better surface finish
  • Includes a certificate of accuracy with NIST traceability
  • Marked with diameter and direction of tolerance for easy identification
  • Suitable for setting micrometers, verniers, and other measuring devices