Learn More
MSC Vermont Gage Plug Gage, Steel, 3.26 mm Diameter, 2 in. Length, 1 ea
Catalog No. 5000514548
Encompass
This Class ZZ minus plug and pin gage is designed for precision inspection and calibration tasks. Manufactured from steel, it provides a measurement diameter of 3.26 mm and an overall length of 2 in. with a tolerance of 0.00508 mm.
  • Measurement diameter of 3.26 mm
  • Overall length of 2 in.
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • Finish of 10 Micro
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, verniers, and other measuring devices, and for use in NoGo inspection
Catalog No. 50-005-14548 Supplier MSC Supplier No. 40430712
This Class ZZ minus plug and pin gage is designed for precision inspection and calibration tasks. Manufactured from steel, it provides a measurement diameter of 3.26 mm and an overall length of 2 in. with a tolerance of 0.00508 mm.
  • Measurement diameter of 3.26 mm
  • Overall length of 2 in.
  • Tolerance of 0.00508 mm
  • Constructed from steel
  • Finish of 10 Micro
  • Includes certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, verniers, and other measuring devices, and for use in NoGo inspection