Learn More
MSC Vermont Gage Plug Gage, Tool Steel, 15.86 mm, 2 in., 1 ea
Catalog No. 5000514350
Encompass
This Class ZZ Plus plug gage is designed for precise measurement applications, including go inspection, setting micrometers, and calibrating verniers or other measuring devices. Constructed from durable tool steel with a hardness of 60/65 Rc, this gage provides long-lasting reliability in demanding environments. It features a 15.86 mm diameter and a 2.0000 in. overall length.
  • Tool steel construction with 60/65 Rc hardness for enhanced durability
  • Finish of 10 microinch or better for smooth surface quality
  • NIST traceable for reliable measurement standards
  • Certificate of accuracy included with each gage
  • Includes marking of size and direction of tolerance
  • Conforms to ASME B89.1 standards
Catalog No. 50-005-14350 Supplier MSC Supplier No. 68106913
This Class ZZ Plus plug gage is designed for precise measurement applications, including go inspection, setting micrometers, and calibrating verniers or other measuring devices. Constructed from durable tool steel with a hardness of 60/65 Rc, this gage provides long-lasting reliability in demanding environments. It features a 15.86 mm diameter and a 2.0000 in. overall length.
  • Tool steel construction with 60/65 Rc hardness for enhanced durability
  • Finish of 10 microinch or better for smooth surface quality
  • NIST traceable for reliable measurement standards
  • Certificate of accuracy included with each gage
  • Includes marking of size and direction of tolerance
  • Conforms to ASME B89.1 standards