Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 9.20 mm, Class ZZ, 1 ea.
Catalog No. 5000514328
Encompass
This high-quality pin gage is designed for precise measurement of hole sizes. Manufactured from durable 52100 tool steel, this gage is heat-treated to 60 to 62 RC for exceptional wear resistance and long-lasting accuracy. It features a 10 micro-finish surface and a length of 5.08 cm (2 in.).
  • Designed for measuring hole sizes with a tolerance of +0.0002 in./0.0000 in.
  • Constructed from 52100 tool steel for high durability and performance
  • Precision surface finish of 10 micro-inches
  • Includes a certificate of accuracy with NIST traceability
Catalog No. 50-005-14328 Supplier MSC Supplier No. 68100254
This high-quality pin gage is designed for precise measurement of hole sizes. Manufactured from durable 52100 tool steel, this gage is heat-treated to 60 to 62 RC for exceptional wear resistance and long-lasting accuracy. It features a 10 micro-finish surface and a length of 5.08 cm (2 in.).
  • Designed for measuring hole sizes with a tolerance of +0.0002 in./0.0000 in.
  • Constructed from 52100 tool steel for high durability and performance
  • Precision surface finish of 10 micro-inches
  • Includes a certificate of accuracy with NIST traceability