Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Plus, 9.19 mm, 1 ea
Catalog No. 5000514327
Encompass
This Class ZZ plus plug gage is a precision measurement tool designed for go inspection, setting micrometers, verniers, and other measuring devices. Constructed from high-quality steel with a 10 microfinish, this member-only reversible pin gage provides reliable performance in industrial metrology applications.
  • Manufactured to ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Diameter of 9.19 mm with a tolerance of 0.00508 mm
  • Overall length of 5.08 cm (2.0000 in.)
  • Reversible style design
  • Made in the USA
Catalog No. 50-005-14327 Supplier MSC Supplier No. 68100247
This Class ZZ plus plug gage is a precision measurement tool designed for go inspection, setting micrometers, verniers, and other measuring devices. Constructed from high-quality steel with a 10 microfinish, this member-only reversible pin gage provides reliable performance in industrial metrology applications.
  • Manufactured to ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
  • Diameter of 9.19 mm with a tolerance of 0.00508 mm
  • Overall length of 5.08 cm (2.0000 in.)
  • Reversible style design
  • Made in the USA