Learn More
MSC Vermont Gage Class ZZ Minus Plug Gage, Steel, 16.72 mm, 2 in. OAL, 1 ea
Catalog No. 5000514156
Encompass
This Class ZZ minus plug and pin gage is designed for precise measurement applications, featuring a 16.72 mm diameter and a 2 in. overall length. It is constructed from durable steel and finished to 10 micro, ensuring high-quality performance in demanding inspection environments.
  • Constructed from steel for durability
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, and calibrating verniers or other measuring devices
  • Tolerance of 0.00508 mm
Catalog No. 50-005-14156 Supplier MSC Supplier No. 40444176
This Class ZZ minus plug and pin gage is designed for precise measurement applications, featuring a 16.72 mm diameter and a 2 in. overall length. It is constructed from durable steel and finished to 10 micro, ensuring high-quality performance in demanding inspection environments.
  • Constructed from steel for durability
  • Includes a certificate of accuracy with NIST traceability
  • Suitable for setting micrometers, NoGo inspection, and calibrating verniers or other measuring devices
  • Tolerance of 0.00508 mm