Learn More
MSC Vermont Gage Plug and Pin Gage, Steel, 16.74 mm, 2 in. Length, 1 ea
Catalog No. 5000514157
Encompass
This Class ZZ minus plug and pin gage is a precision measuring tool with a diameter of 16.74 mm and an overall length of 2 in. Designed for accuracy, it is suitable for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices.
  • Constructed from durable steel with a hardness of 60/65 Rc
  • Features a 10 microinch or better surface finish
  • Includes a certificate of accuracy with NIST traceability
  • Round within 0.0025 mm
  • Marked with size and direction of tolerance
  • Tolerance of 0.00508 mm
Catalog No. 50-005-14157 Supplier MSC Supplier No. 40444192
This Class ZZ minus plug and pin gage is a precision measuring tool with a diameter of 16.74 mm and an overall length of 2 in. Designed for accuracy, it is suitable for setting micrometers, performing NoGo inspections, and calibrating verniers and other measuring devices.
  • Constructed from durable steel with a hardness of 60/65 Rc
  • Features a 10 microinch or better surface finish
  • Includes a certificate of accuracy with NIST traceability
  • Round within 0.0025 mm
  • Marked with size and direction of tolerance
  • Tolerance of 0.00508 mm