Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, 21.64 mm, Tool Steel, 2.0000 in. OAL, 1 ea.
Catalog No. 5000514163
Encompass
This Class ZZ Plus plug gage is designed for precise measurement and inspection tasks, featuring a nominal diameter of 21.64 mm and an overall length of 2.0000 in. Constructed from durable tool steel, it is engineered for reliability in demanding metrology environments.
  • Diameter of 21.64 mm
  • Overall length of 2.0000 in.
  • Tolerance of 0.00508 mm
  • Constructed from tool steel with a 10 micro finish
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards
Catalog No. 50-005-14163 Supplier MSC Supplier No. 68112697
This Class ZZ Plus plug gage is designed for precise measurement and inspection tasks, featuring a nominal diameter of 21.64 mm and an overall length of 2.0000 in. Constructed from durable tool steel, it is engineered for reliability in demanding metrology environments.
  • Diameter of 21.64 mm
  • Overall length of 2.0000 in.
  • Tolerance of 0.00508 mm
  • Constructed from tool steel with a 10 micro finish
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Manufactured to ASME B89.1 standards