Learn More
MSC Vermont Gage Class ZZ Plug Gage, 0.66 mm, Steel, 1 ea.
Catalog No. 5000514078
Encompass
This Class ZZ plug gage is designed for precise measurement and inspection applications, such as calibrating micrometers, verniers, and other measuring devices. It is constructed from durable 52100 tool steel and features a high-precision finish to ensure accurate performance in industrial environments.
  • Constructed from 52100 tool steel, heat-treated to 60 to 62 RC
  • Features a finish of 10 microinch or better for enhanced accuracy
  • NIST traceable and includes a certificate of accuracy
  • Designed for go inspection applications
  • Meets ASME B89.1 standards
Catalog No. 50-005-14078 Supplier MSC Supplier No. 68091719
This Class ZZ plug gage is designed for precise measurement and inspection applications, such as calibrating micrometers, verniers, and other measuring devices. It is constructed from durable 52100 tool steel and features a high-precision finish to ensure accurate performance in industrial environments.
  • Constructed from 52100 tool steel, heat-treated to 60 to 62 RC
  • Features a finish of 10 microinch or better for enhanced accuracy
  • NIST traceable and includes a certificate of accuracy
  • Designed for go inspection applications
  • Meets ASME B89.1 standards