Learn More
MSC Vermont Gage Plug And Pin Gage, Tool Steel, 16.15 mm Diameter, 2 in. Length, 1 ea.
Catalog No. 5000514100
Encompass
This Class ZZ minus plug and pin gage is a precision measurement tool designed for inspecting hole sizes, checking locations, and calibrating measuring devices. Constructed from high-quality tool steel, this gage features a nominal diameter of 16.15 mm and an overall length of 2 in. It is engineered for durability and reliability in demanding shop environments.
  • Hardened to 60 to 65 Rc for extended service life
  • Features a 10 microinch surface finish
  • Includes a certificate of accuracy with NIST traceability
  • Clearly marked with diameter and direction of tolerance
  • Suitable for setting micrometers and verifying other inspection instruments
Catalog No. 50-005-14100 Supplier MSC Supplier No. 40443608
This Class ZZ minus plug and pin gage is a precision measurement tool designed for inspecting hole sizes, checking locations, and calibrating measuring devices. Constructed from high-quality tool steel, this gage features a nominal diameter of 16.15 mm and an overall length of 2 in. It is engineered for durability and reliability in demanding shop environments.
  • Hardened to 60 to 65 Rc for extended service life
  • Features a 10 microinch surface finish
  • Includes a certificate of accuracy with NIST traceability
  • Clearly marked with diameter and direction of tolerance
  • Suitable for setting micrometers and verifying other inspection instruments