Learn More
MSC Vermont Gage Plug Gage, 14.62 mm Diameter, 2 in. Length, 1 ea
Catalog No. 5000514011
Encompass
This Class ZZ minus plug and pin gage is a high-precision measurement tool designed for inspecting, calibrating, and testing applications. Engineered for reliability, this gage provides accurate dimensional verification for micrometers, verniers, and other precision measuring instruments.
  • Manufactured from durable tool steel with a hardness of 60/65 Rc
  • Features a surface finish of 10 microinch or better for improved wear resistance
  • Roundness is accurate within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability
  • Diameter of 14.62 mm with an overall length of 2 in.
  • Tolerance direction specified as minus for NoGo inspection
  • Marked with size and direction of tolerance for quick identification
Catalog No. 50-005-14011 Supplier MSC Supplier No. 40442071
This Class ZZ minus plug and pin gage is a high-precision measurement tool designed for inspecting, calibrating, and testing applications. Engineered for reliability, this gage provides accurate dimensional verification for micrometers, verniers, and other precision measuring instruments.
  • Manufactured from durable tool steel with a hardness of 60/65 Rc
  • Features a surface finish of 10 microinch or better for improved wear resistance
  • Roundness is accurate within 0.0001 in.
  • Includes a certificate of accuracy with NIST traceability
  • Diameter of 14.62 mm with an overall length of 2 in.
  • Tolerance direction specified as minus for NoGo inspection
  • Marked with size and direction of tolerance for quick identification