Learn More
MSC Vermont Gage Class ZZ Pin Gage, Tool Steel, 6.44 mm, 2 in., 1 ea.
Catalog No. 5000513924
Encompass
This Class ZZ Plus pin gage is designed for precision inspection and measurement applications. It is crafted from durable tool steel, featuring a 6.44 mm diameter and an overall length of 2.0000 in. This gage is ideal for Go inspection, setting micrometers, calipers, and snap gages, as well as calculating distances between holes.
  • Manufactured from heat-treated tool steel to 60 to 62 RC
  • Centerless lapped with a 10 microfinish or better
  • Includes a certificate of accuracy with NIST traceability
  • Features ground ends with smooth edges
  • Meets ASME B89.1 standards
Catalog No. 50-005-13924 Supplier MSC Supplier No. 68097492
This Class ZZ Plus pin gage is designed for precision inspection and measurement applications. It is crafted from durable tool steel, featuring a 6.44 mm diameter and an overall length of 2.0000 in. This gage is ideal for Go inspection, setting micrometers, calipers, and snap gages, as well as calculating distances between holes.
  • Manufactured from heat-treated tool steel to 60 to 62 RC
  • Centerless lapped with a 10 microfinish or better
  • Includes a certificate of accuracy with NIST traceability
  • Features ground ends with smooth edges
  • Meets ASME B89.1 standards