Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 5.64 mm, Class ZZ, Minus, 1 ea
Catalog No. 5000513105
Encompass
These Class ZZ pin gages provide a reliable solution for precision inspections and equipment calibration. Engineered for durability, each pin is crafted from tool steel with a hardness of 60/65 Rc and a 10 microinch finish. These gages are ideal for checking hole sizes, measuring slot widths, calculating hole distances, and verifying locations, as well as for use in Go/NoGo gaging and sizing micrometers or snap gages.
  • Individual pin marked with size and direction of tolerance
  • NIST traceable with a certificate of accuracy included on the package
  • Minus tolerance classification for NoGo inspection applications
  • Hardened steel construction for extended wear life
  • Roundness within 0.0001 mm
Catalog No. 50-005-13105 Supplier MSC Supplier No. 40433096
These Class ZZ pin gages provide a reliable solution for precision inspections and equipment calibration. Engineered for durability, each pin is crafted from tool steel with a hardness of 60/65 Rc and a 10 microinch finish. These gages are ideal for checking hole sizes, measuring slot widths, calculating hole distances, and verifying locations, as well as for use in Go/NoGo gaging and sizing micrometers or snap gages.
  • Individual pin marked with size and direction of tolerance
  • NIST traceable with a certificate of accuracy included on the package
  • Minus tolerance classification for NoGo inspection applications
  • Hardened steel construction for extended wear life
  • Roundness within 0.0001 mm