Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Steel, 18.95 mm, 1 ea
Catalog No. 5000513067
Encompass
This metric pin gage is a precision measurement tool designed for go inspection, calibrating micrometers, setting verniers, and verifying other measuring devices. Constructed from high-quality tool steel, this member-only style gage ensures reliable accuracy and durability for diverse industrial metrology applications.
  • Class ZZ tolerance for precise measurement
  • Manufactured from tool steel with a hardness of Rc 60 to 65
  • Features a 10 microfinish or better for smooth operation
  • Includes a certificate of accuracy with NIST traceability
  • Constructed to meet ASME B89.1 standards
Catalog No. 50-005-13067 Supplier MSC Supplier No. 68110006
This metric pin gage is a precision measurement tool designed for go inspection, calibrating micrometers, setting verniers, and verifying other measuring devices. Constructed from high-quality tool steel, this member-only style gage ensures reliable accuracy and durability for diverse industrial metrology applications.
  • Class ZZ tolerance for precise measurement
  • Manufactured from tool steel with a hardness of Rc 60 to 65
  • Features a 10 microfinish or better for smooth operation
  • Includes a certificate of accuracy with NIST traceability
  • Constructed to meet ASME B89.1 standards