Learn More
MSC Vermont Gage Class ZZ Plus Pin Gage, 21.98 mm, Tool Steel, 1 ea
Catalog No. 5000513016
Encompass
This metric pin gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. Constructed from high-quality tool steel, this member-only reversible plug gage ensures durability and reliable performance in metrology applications.
  • Manufactured to ASME B89.1 standards
  • Tolerance of 0.00508 mm
  • Surface finish of 10 micro
  • Overall length of 2.0000 in.
  • Diameter of 21.98 mm
  • Includes certificate of accuracy with NIST traceability
  • Made in the USA
Catalog No. 50-005-13016 Supplier MSC Supplier No. 68113034
This metric pin gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring devices. Constructed from high-quality tool steel, this member-only reversible plug gage ensures durability and reliable performance in metrology applications.
  • Manufactured to ASME B89.1 standards
  • Tolerance of 0.00508 mm
  • Surface finish of 10 micro
  • Overall length of 2.0000 in.
  • Diameter of 21.98 mm
  • Includes certificate of accuracy with NIST traceability
  • Made in the USA