Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 9.69 mm, Plus, 1 ea
Catalog No. 5000512924
Encompass
This Class ZZ Plus plug gage is a precision-manufactured member-only pin gage designed for accurate metrology applications. Constructed from tool steel hardened to 60/65 Rc, this gage provides high durability and reliable performance for tasks such as go inspection, setting micrometers, and calibrating verniers or other measuring devices. The gage features a 9.69 mm diameter and a 2.0000 in. overall length.
  • Features a 10 microinch finish or better for smooth operation
  • Includes a certificate of accuracy with NIST traceability
  • Maintains roundness within 0.0001 in. (0.0025 mm)
  • Tolerance of 0.005 mm (0.0002 in.)
  • Reversible style
  • Meets ASME B89.1 standards
Catalog No. 50-005-12924 Supplier MSC Supplier No. 68100742
This Class ZZ Plus plug gage is a precision-manufactured member-only pin gage designed for accurate metrology applications. Constructed from tool steel hardened to 60/65 Rc, this gage provides high durability and reliable performance for tasks such as go inspection, setting micrometers, and calibrating verniers or other measuring devices. The gage features a 9.69 mm diameter and a 2.0000 in. overall length.
  • Features a 10 microinch finish or better for smooth operation
  • Includes a certificate of accuracy with NIST traceability
  • Maintains roundness within 0.0001 in. (0.0025 mm)
  • Tolerance of 0.005 mm (0.0002 in.)
  • Reversible style
  • Meets ASME B89.1 standards