Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, Tool Steel, 10.29 mm, 2 in. OAL, 1 ea.
Catalog No. 5000512758
Encompass
This Class ZZ Plus plug gage is designed for precision inspection and calibration tasks. Engineered for reliability, this tool features a 10.29 mm diameter and an overall length of 50.8 mm (2.0000 in.).
  • Constructed from durable tool steel
  • Tolerance of +0.00004 in., -0.0000 in.
  • Surface finish of 10 micro
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
Catalog No. 50-005-12758 Supplier MSC Supplier No. 68101344
This Class ZZ Plus plug gage is designed for precision inspection and calibration tasks. Engineered for reliability, this tool features a 10.29 mm diameter and an overall length of 50.8 mm (2.0000 in.).
  • Constructed from durable tool steel
  • Tolerance of +0.00004 in., -0.0000 in.
  • Surface finish of 10 micro
  • Suitable for go inspection and setting micrometers, verniers, and other measuring devices
  • Includes certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards