Learn More
MSC Vermont Gage Pin Gage, Tool Steel, 19.11 mm, 1 ea
Catalog No. 5000512748
Encompass
This Class ZZ plus pin gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring instruments. Constructed from tool steel, it features a reversible style and maintains a diameter of 19.11 mm with an overall length of 2.0000 in.
  • Tolerance of 0.00508 mm
  • Includes certificate of accuracy with NIST traceability
  • Constructed from durable tool steel
  • Meets ASME B89.1 standards
  • Finish of 10 micro
Catalog No. 50-005-12748 Supplier MSC Supplier No. 68110162
This Class ZZ plus pin gage is designed for precise Go inspection and for setting micrometers, verniers, and other measuring instruments. Constructed from tool steel, it features a reversible style and maintains a diameter of 19.11 mm with an overall length of 2.0000 in.
  • Tolerance of 0.00508 mm
  • Includes certificate of accuracy with NIST traceability
  • Constructed from durable tool steel
  • Meets ASME B89.1 standards
  • Finish of 10 micro