Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Plus Tolerance, Tool Steel, 15.29 mm, 2 in., 1 ea
Catalog No. 5000512680
Encompass
This Class ZZ plus plug gage is a high-precision inspection tool designed for go inspection tasks and for setting micrometers, verniers, and other measuring devices. Engineered with a diameter of 15.29 mm and an overall length of 2.0000 in., this member-only reversible gage provides reliable measurements for industrial applications.
  • Constructed from durable tool steel
  • Features a plus direction of tolerance
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Provides a surface finish of 10 micro
Catalog No. 50-005-12680 Supplier MSC Supplier No. 68106343
This Class ZZ plus plug gage is a high-precision inspection tool designed for go inspection tasks and for setting micrometers, verniers, and other measuring devices. Engineered with a diameter of 15.29 mm and an overall length of 2.0000 in., this member-only reversible gage provides reliable measurements for industrial applications.
  • Constructed from durable tool steel
  • Features a plus direction of tolerance
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Provides a surface finish of 10 micro