Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Steel, 15.16 mm, 1 ea.
Catalog No. 5000512589
Encompass
This Class ZZ metric pin gage is designed for precise Go inspection and for setting measuring devices such as micrometers and verniers. Constructed from high-quality tool steel with a hardness of 60/65 Rc, this gage offers a durable and reliable solution for industrial measurement applications.
  • Features a 15.16 mm diameter with a tolerance of 0.00508 mm
  • Constructed from tool steel hardened to 60/65 Rc
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Finish of 10 microinch or better for smooth operation
  • Reversible style
Catalog No. 50-005-12589 Supplier MSC Supplier No. 68106210
This Class ZZ metric pin gage is designed for precise Go inspection and for setting measuring devices such as micrometers and verniers. Constructed from high-quality tool steel with a hardness of 60/65 Rc, this gage offers a durable and reliable solution for industrial measurement applications.
  • Features a 15.16 mm diameter with a tolerance of 0.00508 mm
  • Constructed from tool steel hardened to 60/65 Rc
  • Includes a certificate of accuracy with NIST traceability
  • Meets ASME B89.1 standards
  • Finish of 10 microinch or better for smooth operation
  • Reversible style