Learn More
MSC Vermont Gage Class ZZ Plus Plug Gage, Steel, 16.60 mm, 2.0000 in. OAL, 1 ea.
Catalog No. 5000512591
Encompass
This Class ZZ plug gage is designed for high-precision go inspection and the calibration of micrometers, verniers, and other measuring instruments. It is constructed from durable tool steel and features a reversible design for extended tool life.
  • Diameter of 16.60 mm
  • Overall length of 2.0000 in.
  • Tolerance of 0.00508 mm
  • Class ZZ tolerance rating
  • Plus tolerance direction
  • Meets ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability
Catalog No. 50-005-12591 Supplier MSC Supplier No. 68107655
This Class ZZ plug gage is designed for high-precision go inspection and the calibration of micrometers, verniers, and other measuring instruments. It is constructed from durable tool steel and features a reversible design for extended tool life.
  • Diameter of 16.60 mm
  • Overall length of 2.0000 in.
  • Tolerance of 0.00508 mm
  • Class ZZ tolerance rating
  • Plus tolerance direction
  • Meets ASME B89.1 standards
  • Includes certificate of accuracy with NIST traceability