Learn More
MSC Vermont Gage Pin Gage, Class ZZ, Minus, 11.35 mm, 1 ea
Catalog No. 5000512537
Encompass
This Class ZZ pin gage is a precision-machined tool designed for high-accuracy inspection and calibration applications. Featuring a 11.35 mm diameter and a 2 in. overall length, this gage is constructed from durable tool steel with a 10 microfinish or better, ensuring reliable performance in demanding measurement environments.
  • Designed for applications including setting micrometers, verniers, and other measuring devices
  • No-go (minus) tolerance type for accurate inspection
  • Constructed from tool steel with a 10 microfinish for durability
  • Includes a certificate of accuracy with NIST traceability
  • Made in the United States
Catalog No. 50-005-12537 Supplier MSC Supplier No. 40438806
This Class ZZ pin gage is a precision-machined tool designed for high-accuracy inspection and calibration applications. Featuring a 11.35 mm diameter and a 2 in. overall length, this gage is constructed from durable tool steel with a 10 microfinish or better, ensuring reliable performance in demanding measurement environments.
  • Designed for applications including setting micrometers, verniers, and other measuring devices
  • No-go (minus) tolerance type for accurate inspection
  • Constructed from tool steel with a 10 microfinish for durability
  • Includes a certificate of accuracy with NIST traceability
  • Made in the United States