Learn More
MSC Vermont Gage Class ZZ Pin Gage, 2.10 mm Diameter, Tool Steel, Plus Tolerance, 1 ea
Catalog No. 5000512252
Encompass
This Class ZZ plug gage is designed for precise measurement applications, including go inspection and the calibration of micrometers, verniers, and other measuring equipment. The gage is constructed from 52100 tool steel and is heat-treated to a hardness of 60 to 62 RC for durability and wear resistance.
  • Diameter of 2.10 mm with a plus tolerance of 0.00508 mm
  • Overall length of 2.0000 in.
  • Finished to a surface roughness of 10 micro or better
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from 52100 tool steel
  • Complies with ASME B89.1 standards
Catalog No. 50-005-12252 Supplier MSC Supplier No. 68093152
This Class ZZ plug gage is designed for precise measurement applications, including go inspection and the calibration of micrometers, verniers, and other measuring equipment. The gage is constructed from 52100 tool steel and is heat-treated to a hardness of 60 to 62 RC for durability and wear resistance.
  • Diameter of 2.10 mm with a plus tolerance of 0.00508 mm
  • Overall length of 2.0000 in.
  • Finished to a surface roughness of 10 micro or better
  • Includes a certificate of accuracy with NIST traceability
  • Constructed from 52100 tool steel
  • Complies with ASME B89.1 standards