Learn More
MSC Vermont Gage Pin Gage, 2.33 mm, Tool Steel, 1 ea.
Catalog No. 5000512091
Encompass
This Class ZZ plus pin gage is designed for high-precision measurement tasks, including Go inspection and the calibration of micrometers, verniers, and other measuring instruments. Constructed from durable tool steel, this reversible gage provides reliable performance in industrial metrology applications.
  • Manufactured to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish for enhanced measurement precision
  • Reversible design for extended service life
  • Tolerance of 0.00508 mm
Catalog No. 50-005-12091 Supplier MSC Supplier No. 68093384
This Class ZZ plus pin gage is designed for high-precision measurement tasks, including Go inspection and the calibration of micrometers, verniers, and other measuring instruments. Constructed from durable tool steel, this reversible gage provides reliable performance in industrial metrology applications.
  • Manufactured to ASME B89.1 standards
  • Includes a certificate of accuracy with NIST traceability
  • Features a 10 micro finish for enhanced measurement precision
  • Reversible design for extended service life
  • Tolerance of 0.00508 mm