Learn More
Medchemexpress LLC HY-134955 5mg , VT103 CAS:2290608-13-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1349555MG
Encompass_Preferred
HY-134955 5mg VT103 CAS: 2290608-13-6 VT103, an analog of VT101, is an orally active and selective TEAD1 protein palmitoylation inhibitor. VT103 inhibits YAP/TAZ-TEAD promoted gene transcription, blocks TEAD auto-palmitoylation, and disrupts interaction between YAP/TAZ and TEAD. VT103 can be used for the research of cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9011