Learn More
Mti Corporation VTC 100A COMPAC SPN COATR 110V
SDP
Catalog No. NC1507856
Encompass
Compact Spin Coater ( Max. 8000 rpm, 4" wafer Max.) with 3 Sets of Vacuum Chucks & Complete Accessories, 110VVTC-100A-110V is CE certified compact spin coater with vacuum chucks designed for easy and quick coating via sol-gel for wafers up to 4". The spinning speed is variable from 500-8000 rpm with two programmable segments. The package comes included with complete accessories for immediate use
Catalog No. NC1507856 Supplier Mti Corporation Supplier No. VTC100AMACH
Compact Spin Coater ( Max. 8000 rpm, 4" wafer Max.) with 3 Sets of Vacuum Chucks & Complete Accessories, 110VVTC-100A-110V is CE certified compact spin coater with vacuum chucks designed for easy and quick coating via sol-gel for wafers up to 4". The spinning speed is variable from 500-8000 rpm with two programmable segments. The package comes included with complete accessories for immediate use