Learn More
Labnet International, Inc VorTemp™ 1550 Incubator/Shaker, Complete with racks for 50 ml and 15 ml tubes, 115V
Catalog No. NC1419694
Encompass_Preferred
Designed for simultaneous heating and mixing of small samples, the VorTemp™ 1550 shaking incubators are supplied with interchangeable platforms for standard 15ml and 50ml conical tubes. Maximum capacity is 8 x 50ml or 18 x 15ml. Temperature in the VorTemp™ 1550 is adjustable over a broad range, making the unit useful for a variety of applications, including bacterial cultures and digestions. Mechanical convection provides a uniform and stable environment within the chamber. The VorTemp™ 1550 may also be used in the cold room for sub-ambient applications.
Catalog No. NC1419694 Supplier Labnet International, Inc Supplier No. S2050A
Designed for simultaneous heating and mixing of small samples, the VorTemp™ 1550 shaking incubators are supplied with interchangeable platforms for standard 15ml and 50ml conical tubes. Maximum capacity is 8 x 50ml or 18 x 15ml. Temperature in the VorTemp™ 1550 is adjustable over a broad range, making the unit useful for a variety of applications, including bacterial cultures and digestions. Mechanical convection provides a uniform and stable environment within the chamber. The VorTemp™ 1550 may also be used in the cold room for sub-ambient applications.