Learn More
Aviva Systems Biology Corp VTI1A Antibody - N-terminal region (ARP52812_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52812_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against VTI1A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human VTI1A. Alternate names: MVti1, Vti1-rp2, VTI1RP2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-60839